Learn More
Chemglass Life Sciences Chemglass O-Ring #014, Simriz 486 FFKM, White, USP Class VI
SDP
Catalog No. 502743529
Encompass_Preferred
Chemglass O-Ring #014 (CG-309-Z-014) is a white perfluoroelastomer (FFKM) sealing element formulated with Simriz 486 and certified to USP Class VI. It is designed for demanding, chemically aggressive dry process environments, including reactive plasmas, where broad chemical resistance and purity are required.
  • Perfluoroelastomer (FFKM) construction for broad chemical resistance.
  • Certified to USP Class VI.
  • White color for easy visual inspection.
  • Suitable for aggressive dry process chemistries, including reactive plasmas.
  • Intended for use in high-purity and vacuum process sealing applications.
Catalog No. 50-274-3529 Supplier Chemglass Life Sciences Supplier No. CG309Z014
Chemglass O-Ring #014 (CG-309-Z-014) is a white perfluoroelastomer (FFKM) sealing element formulated with Simriz 486 and certified to USP Class VI. It is designed for demanding, chemically aggressive dry process environments, including reactive plasmas, where broad chemical resistance and purity are required.
  • Perfluoroelastomer (FFKM) construction for broad chemical resistance.
  • Certified to USP Class VI.
  • White color for easy visual inspection.
  • Suitable for aggressive dry process chemistries, including reactive plasmas.
  • Intended for use in high-purity and vacuum process sealing applications.