Learn More
Chemglass Life Sciences Chemglass O-Ring CG-309-Z-015, Simriz 486 FFKM (Size #-015)
SDP
Catalog No. 502743528
Encompass_Preferred
The Chemglass CG-309-Z-015 O-ring is a perfluoroelastomer (FFKM) sealing component manufactured from Simriz 486 for high-purity, high-temperature laboratory and process applications.
  • Perfluoroelastomer material with broad chemical resistance.
  • Certified to USP Class VI for biocompatibility.
  • White color and 75 Shore A hardness.
  • Service temperature range of -7°C to 230°C.
  • Tensile strength approximately 2,620 psi.
  • Suitable for aggressive chemistries, reactive plasmas, valves, and fittings.
  • Manufactured in clean-room conditions for high purity.
Catalog No. 50-274-3528 Supplier Chemglass Life Sciences Supplier No. CG309Z015
The Chemglass CG-309-Z-015 O-ring is a perfluoroelastomer (FFKM) sealing component manufactured from Simriz 486 for high-purity, high-temperature laboratory and process applications.
  • Perfluoroelastomer material with broad chemical resistance.
  • Certified to USP Class VI for biocompatibility.
  • White color and 75 Shore A hardness.
  • Service temperature range of -7°C to 230°C.
  • Tensile strength approximately 2,620 psi.
  • Suitable for aggressive chemistries, reactive plasmas, valves, and fittings.
  • Manufactured in clean-room conditions for high purity.