Learn More
Chemglass Life Sciences Chemglass O-Ring, AS568 #-016 - Simriz 486 FFKM perfluoro, white; for vacuum joints and laboratory glassware
SDP
Catalog No. 502743527
Encompass_Preferred
Chemglass O-Ring CG-309-Z-016 is an AS568 #-016 size perfluoro (FFKM) O-ring molded from Simriz 486 material and supplied in white with Class VI cleanliness. It is intended for reliable sealing in laboratory glassware and vacuum joint applications, including use in demanding chemical and semiconductor environments.
  • Provides broad chemical resistance.
  • Offers high temperature stability for demanding processes.
  • White color supports visual cleanliness checks.
  • Suitable for vacuum joints and laboratory glassware seals.
  • Class VI cleanliness suitable for pharmaceutical or semiconductor use.
Catalog No. 50-274-3527 Supplier Chemglass Life Sciences Supplier No. CG309Z016
Chemglass O-Ring CG-309-Z-016 is an AS568 #-016 size perfluoro (FFKM) O-ring molded from Simriz 486 material and supplied in white with Class VI cleanliness. It is intended for reliable sealing in laboratory glassware and vacuum joint applications, including use in demanding chemical and semiconductor environments.
  • Provides broad chemical resistance.
  • Offers high temperature stability for demanding processes.
  • White color supports visual cleanliness checks.
  • Suitable for vacuum joints and laboratory glassware seals.
  • Class VI cleanliness suitable for pharmaceutical or semiconductor use.