Learn More
Chemglass Life Sciences Chemglass O-Ring #019, Simriz 486 FFKM Perfluoroelastomer, White, Class VI
SDP
Catalog No. 502743525
Encompass_Preferred
This high-purity perfluoroelastomer O-ring (size #019) is designed for demanding laboratory sealing applications where broad chemical resistance and high temperature stability are required. It is formulated from a perfluoroelastomer compound and supplied in a white, Class VI material.
  • Material: perfluoroelastomer (FFKM)
  • Color: white
  • Durometer: 75 (unless otherwise noted)
  • Effective service temperature range: -7 to 230° C
  • Suitable for acids, inorganic bases, alcohols/ethers, hydrocarbons, esters, aldehydes/ketones, amines, and chlorocarbon solvents
  • Common uses: laboratory O-ring joints and vacuum traps
  • Certification: Class VI biocompatibility (as listed for the series)
Catalog No. 50-274-3525 Supplier Chemglass Life Sciences Supplier No. CG309Z019
This high-purity perfluoroelastomer O-ring (size #019) is designed for demanding laboratory sealing applications where broad chemical resistance and high temperature stability are required. It is formulated from a perfluoroelastomer compound and supplied in a white, Class VI material.
  • Material: perfluoroelastomer (FFKM)
  • Color: white
  • Durometer: 75 (unless otherwise noted)
  • Effective service temperature range: -7 to 230° C
  • Suitable for acids, inorganic bases, alcohols/ethers, hydrocarbons, esters, aldehydes/ketones, amines, and chlorocarbon solvents
  • Common uses: laboratory O-ring joints and vacuum traps
  • Certification: Class VI biocompatibility (as listed for the series)