Learn More
Chemglass Life Sciences Chemglass O-Ring #020, Simriz 486 FFKM (Perfluoro), White, USP Class VI
SDP
Catalog No. 502743524
Encompass_Preferred
Chemglass O-Ring #020 is a high-purity perfluoroelastomer (FFKM) sealing ring made from Simriz 486, engineered for broad chemical resistance and high-temperature service. The white, USP Class VI material combines elastomer resilience with chemical resistance approaching PTFE, making it suitable for aggressive process chemistries and reactive plasma environments.
  • Perfluoroelastomer (FFKM) construction for broad chemical resistance.
  • White, high-purity material certified to USP Class VI.
  • Suitable for aggressive dry process chemistries and reactive plasmas.
  • Effective service temperature up to 220 °C.
  • Size #020 configured for standard O-ring fittings.
Catalog No. 50-274-3524 Supplier Chemglass Life Sciences Supplier No. CG309Z020
Chemglass O-Ring #020 is a high-purity perfluoroelastomer (FFKM) sealing ring made from Simriz 486, engineered for broad chemical resistance and high-temperature service. The white, USP Class VI material combines elastomer resilience with chemical resistance approaching PTFE, making it suitable for aggressive process chemistries and reactive plasma environments.
  • Perfluoroelastomer (FFKM) construction for broad chemical resistance.
  • White, high-purity material certified to USP Class VI.
  • Suitable for aggressive dry process chemistries and reactive plasmas.
  • Effective service temperature up to 220 °C.
  • Size #020 configured for standard O-ring fittings.