Learn More
Chemglass Life Sciences Chemglass O-Ring CG-309-Z-021 - SIMRIZ 486 FFKM perfluoro, white, size 021
SDP
Catalog No. 502743523
Encompass_Preferred
This Chemglass O-ring (CG-309-Z-021) is a perfluoroelastomer sealing element made from SIMRIZ 486 FFKM, engineered for broad chemical compatibility and thermal stability for laboratory vacuum joints and fittings.
  • Perfluoroelastomer material for broad chemical resistance.
  • White color with Class VI biocompatibility.
  • Typically stocked at 75 durometer hardness.
  • Designed for use in vacuum joints and laboratory fittings.
  • Suitable for high temperature and aggressive chemical environments.
Catalog No. 50-274-3523 Supplier Chemglass Life Sciences Supplier No. CG309Z021
This Chemglass O-ring (CG-309-Z-021) is a perfluoroelastomer sealing element made from SIMRIZ 486 FFKM, engineered for broad chemical compatibility and thermal stability for laboratory vacuum joints and fittings.
  • Perfluoroelastomer material for broad chemical resistance.
  • White color with Class VI biocompatibility.
  • Typically stocked at 75 durometer hardness.
  • Designed for use in vacuum joints and laboratory fittings.
  • Suitable for high temperature and aggressive chemical environments.