Learn More
Chemglass Life Sciences Simriz 486 FFKM O-ring, AS568-022 - 0.989 in (25.12 mm) ID x 0.070 in (1.78 mm) CS, white, 75 Shore A
SDP
Catalog No. 502743522
Encompass_Preferred
Simriz 486 FFKM O-ring (AS568-022) is a white perfluoroelastomer seal designed for ultra-clean and chemically aggressive environments. It provides high thermal stability, excellent chemical and plasma resistance, and low particulate generation for semiconductor and pharmaceutical applications.
  • High chemical and temperature resistance.
  • Low particulation for clean processing environments.
  • White, carbon-black free formulation suitable for pharmaceutical and semiconductor use.
  • 75 Shore A hardness for resilient sealing performance.
  • Standard AS568 profile for interchangeability and easy replacement.
Catalog No. 50-274-3522 Supplier Chemglass Life Sciences Supplier No. CG309Z022
Simriz 486 FFKM O-ring (AS568-022) is a white perfluoroelastomer seal designed for ultra-clean and chemically aggressive environments. It provides high thermal stability, excellent chemical and plasma resistance, and low particulate generation for semiconductor and pharmaceutical applications.
  • High chemical and temperature resistance.
  • Low particulation for clean processing environments.
  • White, carbon-black free formulation suitable for pharmaceutical and semiconductor use.
  • 75 Shore A hardness for resilient sealing performance.
  • Standard AS568 profile for interchangeability and easy replacement.