Learn More
Chemglass Life Sciences Perfluoro (FFKM) O-ring, Simriz 486 compound, size #024 - for vacuum traps and O-ring joints
SDP
Catalog No. 502743520
Encompass_Preferred
Chemglass perfluoro O-ring manufactured in Simriz 486 FFKM compound, presented in a white, pharmaceutical-grade formulation suitable for laboratory glassware seals and vacuum applications.
  • Perfluoroelastomer (FFKM) material with broad chemical resistance.
  • White color, typically 75 durometer.
  • Class VI compatible when manufactured from the Simriz 486 variant.
  • Designed for vacuum traps and O-ring joints.
  • Dash size #024 for standard O-ring sizing.
Catalog No. 50-274-3520 Supplier Chemglass Life Sciences Supplier No. CG309Z024
Chemglass perfluoro O-ring manufactured in Simriz 486 FFKM compound, presented in a white, pharmaceutical-grade formulation suitable for laboratory glassware seals and vacuum applications.
  • Perfluoroelastomer (FFKM) material with broad chemical resistance.
  • White color, typically 75 durometer.
  • Class VI compatible when manufactured from the Simriz 486 variant.
  • Designed for vacuum traps and O-ring joints.
  • Dash size #024 for standard O-ring sizing.