Learn More
Chemglass Life Sciences Chemglass O-Ring CG-309-Z-026 - Simriz 486 FFKM White, Size 026 (for Vacuum Traps and O-Ring Joints)
SDP
Catalog No. 502743519
Encompass_Preferred
The CG-309-Z-026 O-ring is a white perfluoroelastomer (FFKM) seal made from Simriz 486, designed to provide high chemical and thermal resistance for laboratory vacuum traps, O-ring joints, and demanding process sealing applications. It meets Class VI quality standards and has an effective service temperature range of -7 to 230 °C.
  • White perfluoroelastomer (FFKM) material.
  • Class VI quality for pharmaceutical and high-purity applications.
  • Effective service temperature: -7 to 230 °C.
  • Broad chemical resistance to acids, inorganic bases, alcohols/ethers, aromatic hydrocarbons, esters, aldehydes/ketones, aliphatic hydrocarbons, amines, and chlorocarbon solvents.
  • Suitable for vacuum traps, O-ring joints, and process sealing applications.
  • Available in AS568 size 026.
Catalog No. 50-274-3519 Supplier Chemglass Life Sciences Supplier No. CG309Z026
The CG-309-Z-026 O-ring is a white perfluoroelastomer (FFKM) seal made from Simriz 486, designed to provide high chemical and thermal resistance for laboratory vacuum traps, O-ring joints, and demanding process sealing applications. It meets Class VI quality standards and has an effective service temperature range of -7 to 230 °C.
  • White perfluoroelastomer (FFKM) material.
  • Class VI quality for pharmaceutical and high-purity applications.
  • Effective service temperature: -7 to 230 °C.
  • Broad chemical resistance to acids, inorganic bases, alcohols/ethers, aromatic hydrocarbons, esters, aldehydes/ketones, aliphatic hydrocarbons, amines, and chlorocarbon solvents.
  • Suitable for vacuum traps, O-ring joints, and process sealing applications.
  • Available in AS568 size 026.