Learn More
Chemglass Life Sciences Chemglass O-Ring, #027 - Perfluoroelastomer (FFKM), White, USP Class VI (CG-309-Z-027)
SDP
Catalog No. 502743518
Encompass_Preferred
Chemglass CG-309-Z-027 is a white perfluoroelastomer (FFKM) O-ring from the CG-309 product family, manufactured using Simriz 486 material and classified to USP Class VI. It provides a high-purity sealing solution for laboratory and process applications.
  • Perfluoroelastomer (FFKM) material
  • White color
  • USP Class VI biocompatibility
  • O-ring size #027
  • Suitable for high-purity and laboratory sealing applications
Catalog No. 50-274-3518 Supplier Chemglass Life Sciences Supplier No. CG309Z027
Chemglass CG-309-Z-027 is a white perfluoroelastomer (FFKM) O-ring from the CG-309 product family, manufactured using Simriz 486 material and classified to USP Class VI. It provides a high-purity sealing solution for laboratory and process applications.
  • Perfluoroelastomer (FFKM) material
  • White color
  • USP Class VI biocompatibility
  • O-ring size #027
  • Suitable for high-purity and laboratory sealing applications