Learn More
Chemglass Life Sciences O-ring, #030, FFKM (perfluoroelastomer), white - replacement sealing O-ring for laboratory glassware and fluid fittings
SDP
Catalog No. 502743517
Encompass_Preferred
O-ring, size #030, made from perfluoroelastomer (FFKM) for chemical resistance and Class VI cleanliness. Designed as a replacement sealing component for laboratory glassware and fluid handling connections.
  • Perfluoroelastomer construction provides broad chemical and temperature resistance.
  • White color enables visual inspection for contamination or wear.
  • Class VI cleanliness suitable for laboratory and medical applications.
  • Standard #030 size for common sealing joints and fittings.
  • Compatible with a wide range of aggressive chemicals.
Catalog No. 50-274-3517 Supplier Chemglass Life Sciences Supplier No. CG309Z030
O-ring, size #030, made from perfluoroelastomer (FFKM) for chemical resistance and Class VI cleanliness. Designed as a replacement sealing component for laboratory glassware and fluid handling connections.
  • Perfluoroelastomer construction provides broad chemical and temperature resistance.
  • White color enables visual inspection for contamination or wear.
  • Class VI cleanliness suitable for laboratory and medical applications.
  • Standard #030 size for common sealing joints and fittings.
  • Compatible with a wide range of aggressive chemicals.