Learn More
Chemglass Life Sciences O-ring, Perfluoro (FFKM), Simriz 486, size #031 - Chemglass CG-309-Z-031
SDP
Catalog No. 502743516
Encompass_Preferred
Perfluoro (FFKM) O-ring fabricated from Simriz 486 material for high-purity sealing in laboratory, pharmaceutical, and semiconductor equipment. The white perfluoroelastomer provides broad chemical compatibility and high temperature stability for aggressive process chemistries.
  • High chemical resistance across a wide range of solvents and acids.
  • Rated for continuous service up to 220 °C.
  • Manufactured in certified clean-room facilities.
  • White, high-purity compound suitable for low-contamination applications.
  • Suitable as a replacement seal for glassware, valves, and process equipment.
Catalog No. 50-274-3516 Supplier Chemglass Life Sciences Supplier No. CG309Z031
Perfluoro (FFKM) O-ring fabricated from Simriz 486 material for high-purity sealing in laboratory, pharmaceutical, and semiconductor equipment. The white perfluoroelastomer provides broad chemical compatibility and high temperature stability for aggressive process chemistries.
  • High chemical resistance across a wide range of solvents and acids.
  • Rated for continuous service up to 220 °C.
  • Manufactured in certified clean-room facilities.
  • White, high-purity compound suitable for low-contamination applications.
  • Suitable as a replacement seal for glassware, valves, and process equipment.