Learn More
Chemglass Life Sciences O-Ring #032, Simriz 486 FFKM - Perfluoro O-Ring for Laboratory Seals
SDP
Catalog No. 502743515
Encompass_Preferred
Perfluoro (FFKM) O-ring manufactured from Simriz 486 material, designed for chemical resistance and high-temperature sealing in laboratory glassware and fittings.
  • Perfluoroelastomer construction for exceptional chemical resistance.
  • Suitable for high-temperature applications and aggressive solvents.
  • White color aids visual inspection and identification.
  • Meets Class VI standards for biocompatibility where applicable.
  • Designed for use with laboratory glassware fittings and assemblies.
Catalog No. 50-274-3515 Supplier Chemglass Life Sciences Supplier No. CG309Z032
Perfluoro (FFKM) O-ring manufactured from Simriz 486 material, designed for chemical resistance and high-temperature sealing in laboratory glassware and fittings.
  • Perfluoroelastomer construction for exceptional chemical resistance.
  • Suitable for high-temperature applications and aggressive solvents.
  • White color aids visual inspection and identification.
  • Meets Class VI standards for biocompatibility where applicable.
  • Designed for use with laboratory glassware fittings and assemblies.