Learn More
Chemglass Life Sciences Chemglass O-Ring #034 - Simriz 486 FFKM perfluoro O-ring, white, USP Class VI - Replacement seal for laboratory and process equipment
SDP
Catalog No. 502743514
Encompass_Preferred
Chemglass O-Ring #034 is a white perfluoroelastomer (FFKM) seal made from Simriz 486. It provides broad chemical resistance, USP Class VI cleanliness, and elevated temperature performance for demanding laboratory, pharmaceutical, and semiconductor sealing applications.
  • Perfluoroelastomer (FFKM) material with USP Class VI certification.
  • White color.
  • Hardness 75 Shore A.
  • Temperature range -7°C to +230°C.
  • Tensile strength 2,620 psi.
  • Supplied individually (each).
Catalog No. 50-274-3514 Supplier Chemglass Life Sciences Supplier No. CG309Z034
Chemglass O-Ring #034 is a white perfluoroelastomer (FFKM) seal made from Simriz 486. It provides broad chemical resistance, USP Class VI cleanliness, and elevated temperature performance for demanding laboratory, pharmaceutical, and semiconductor sealing applications.
  • Perfluoroelastomer (FFKM) material with USP Class VI certification.
  • White color.
  • Hardness 75 Shore A.
  • Temperature range -7°C to +230°C.
  • Tensile strength 2,620 psi.
  • Supplied individually (each).