Learn More
Agilent Technologies O-ring, 1/32 inch id, 3/32 inch od
Catalog No. NC1805182
Encompass
O-ring, 1/32 inch id, 3/32 inch od, 1/32 inch nitrile, for capillary assembly in Mark 7 spray chamber.
Catalog No. NC1805182 Supplier Agilent Technologies Supplier No. 6910009700
O-ring, 1/32 inch id, 3/32 inch od, 1/32 inch nitrile, for capillary assembly in Mark 7 spray chamber.