Learn More
Chemglass Life Sciences Chemglass O-Ring #104, Simriz 486 FFKM, White (CG-309-Z-104)
SDP
Catalog No. 502743513
Encompass_Preferred
Chemglass O-ring #104 (CG-309-Z-104) made from Simriz 486 perfluoroelastomer (FFKM) provides high chemical resistance and thermal stability for demanding laboratory, pharmaceutical, and semiconductor sealing applications. The white, Class VI material is suitable for cleanroom and biocompatible uses and fits standard size 104 O-ring grooves.
  • High chemical resistance suitable for aggressive solvents and acids.
  • High temperature stability for demanding thermal environments.
  • White color aids contamination control and visual inspection.
  • Class VI biocompatibility for pharmaceutical and medical applications.
  • Standard size #104 for easy replacement and compatibility with common O-ring fittings.
Catalog No. 50-274-3513 Supplier Chemglass Life Sciences Supplier No. CG309Z104
Chemglass O-ring #104 (CG-309-Z-104) made from Simriz 486 perfluoroelastomer (FFKM) provides high chemical resistance and thermal stability for demanding laboratory, pharmaceutical, and semiconductor sealing applications. The white, Class VI material is suitable for cleanroom and biocompatible uses and fits standard size 104 O-ring grooves.
  • High chemical resistance suitable for aggressive solvents and acids.
  • High temperature stability for demanding thermal environments.
  • White color aids contamination control and visual inspection.
  • Class VI biocompatibility for pharmaceutical and medical applications.
  • Standard size #104 for easy replacement and compatibility with common O-ring fittings.