Learn More
Chemglass Life Sciences Chemglass O-Ring, Simriz 486 FFKM, #105 (CG-309-Z-105)
SDP
Catalog No. 502743512
Encompass_Preferred
Perfluoroelastomer O-ring molded from Simriz 486 FFKM, designed for high-purity sealing in laboratory, pharmaceutical, and semiconductor equipment. The white perfluoroelastomer provides broad chemical resistance and high temperature stability, and is indicated as meeting USP Class VI biocompatibility by the manufacturer.
  • Perfluoroelastomer (FFKM) construction for chemical resistance.
  • High temperature stability for process and laboratory use.
  • White color suited for cleanroom environments and visual inspection.
  • USP Class VI biocompatible where specified.
  • Sized to O-ring #105 for direct replacement.
Catalog No. 50-274-3512 Supplier Chemglass Life Sciences Supplier No. CG309Z105
Perfluoroelastomer O-ring molded from Simriz 486 FFKM, designed for high-purity sealing in laboratory, pharmaceutical, and semiconductor equipment. The white perfluoroelastomer provides broad chemical resistance and high temperature stability, and is indicated as meeting USP Class VI biocompatibility by the manufacturer.
  • Perfluoroelastomer (FFKM) construction for chemical resistance.
  • High temperature stability for process and laboratory use.
  • White color suited for cleanroom environments and visual inspection.
  • USP Class VI biocompatible where specified.
  • Sized to O-ring #105 for direct replacement.