Learn More
Chemglass Life Sciences Chemglass O-Ring #107, Simriz 486 FFKM for laboratory glassware joints (CG-309-Z-107)
SDP
Catalog No. 502743511
Encompass_Preferred
Chemglass CG-309-Z-107 is a perfluoroelastomer (FFKM) O-ring made from Simriz 486 material and sized to AS568 #107. It is supplied in a white, Class VI formulation for high-purity laboratory sealing applications, and is intended for use in glassware joints, vacuum traps, and chemical processing fittings.
  • High chemical resistance due to Simriz 486 FFKM material.
  • White, high-purity formulation suitable for laboratory use.
  • Class VI cleanliness for applications requiring biocompatibility and low extractables.
  • Sized to AS568-107 (size #107) for compatibility with common lab fittings.
  • Designed for sealing glassware joints and vacuum systems in laboratory environments.
Catalog No. 50-274-3511 Supplier Chemglass Life Sciences Supplier No. CG309Z107
Chemglass CG-309-Z-107 is a perfluoroelastomer (FFKM) O-ring made from Simriz 486 material and sized to AS568 #107. It is supplied in a white, Class VI formulation for high-purity laboratory sealing applications, and is intended for use in glassware joints, vacuum traps, and chemical processing fittings.
  • High chemical resistance due to Simriz 486 FFKM material.
  • White, high-purity formulation suitable for laboratory use.
  • Class VI cleanliness for applications requiring biocompatibility and low extractables.
  • Sized to AS568-107 (size #107) for compatibility with common lab fittings.
  • Designed for sealing glassware joints and vacuum systems in laboratory environments.