Learn More
Chemglass Life Sciences Chemglass Perfluoro O-Ring (FFKM), Simriz 486, Size #108 - For O-Ring Joints and Vacuum Traps
SDP
Catalog No. 502743510
Encompass_Preferred
Chemglass perfluoro O-rings (CG-309-Z series) use a high-purity Simriz 486 FFKM compound to provide broad chemical compatibility and thermal stability for laboratory sealing applications. Size #108 is stocked as a white, Class VI-capable perfluoroelastomer O-ring suitable for O-ring joints and vacuum traps.
  • Perfluoroelastomer (FFKM) compound for broad chemical resistance.
  • White color and 75 durometer for general laboratory sealing.
  • Effective service temperature -7 °C to 230 °C.
  • Suitable for vacuum traps and O-ring joints.
  • Available in standard laboratory O-ring sizes; size shown in product listing.
Catalog No. 50-274-3510 Supplier Chemglass Life Sciences Supplier No. CG309Z108
Chemglass perfluoro O-rings (CG-309-Z series) use a high-purity Simriz 486 FFKM compound to provide broad chemical compatibility and thermal stability for laboratory sealing applications. Size #108 is stocked as a white, Class VI-capable perfluoroelastomer O-ring suitable for O-ring joints and vacuum traps.
  • Perfluoroelastomer (FFKM) compound for broad chemical resistance.
  • White color and 75 durometer for general laboratory sealing.
  • Effective service temperature -7 °C to 230 °C.
  • Suitable for vacuum traps and O-ring joints.
  • Available in standard laboratory O-ring sizes; size shown in product listing.