Learn More
Chemglass Life Sciences O-ring AS568-109, Simriz 486 FFKM - 7.595 x 2.616 mm (0.299 x 0.103 in)
SDP
Catalog No. 502743509
Encompass_Preferred
O-ring manufactured in perfluoroelastomer (FFKM) for demanding sealing applications. The o-ring offers chemical resistance and thermal stability typical of FFKM elastomers, and is provided in a white finish for easy inspection.
  • Perfluoroelastomer construction for broad chemical resistance.
  • High-temperature stability suitable for demanding environments.
  • Precision molded for consistent dimensional tolerances.
  • White finish for visual inspection and contamination control.
  • Compact cross-section for sealing small-diameter components.
Catalog No. 50-274-3509 Supplier Chemglass Life Sciences Supplier No. CG309Z109
O-ring manufactured in perfluoroelastomer (FFKM) for demanding sealing applications. The o-ring offers chemical resistance and thermal stability typical of FFKM elastomers, and is provided in a white finish for easy inspection.
  • Perfluoroelastomer construction for broad chemical resistance.
  • High-temperature stability suitable for demanding environments.
  • Precision molded for consistent dimensional tolerances.
  • White finish for visual inspection and contamination control.
  • Compact cross-section for sealing small-diameter components.