Learn More
Chemglass Life Sciences O-Ring AS568-110 (Simriz 485 FFKM perfluoroelastomer) for Reaction Vessels and Lids
SDP
Catalog No. 502743603
Encompass_Preferred
Chemglass O-ring CG-309-B-110 is an AS568-110 size perfluoroelastomer (FFKM) sealing ring made from Simriz 485 material. It provides broad chemical resistance and elevated-temperature sealing for laboratory reaction vessels, reactors, and lids.
  • Perfluoroelastomer (FFKM) material for broad chemical resistance.
  • AS568-110 size for standard O-ring grooves.
  • Black color for visual identification.
  • Suitable for high-temperature laboratory sealing applications.
  • Used to seal reaction vessels and lids.
Catalog No. 50-274-3603 Supplier Chemglass Life Sciences Supplier No. CG309B110
Chemglass O-ring CG-309-B-110 is an AS568-110 size perfluoroelastomer (FFKM) sealing ring made from Simriz 485 material. It provides broad chemical resistance and elevated-temperature sealing for laboratory reaction vessels, reactors, and lids.
  • Perfluoroelastomer (FFKM) material for broad chemical resistance.
  • AS568-110 size for standard O-ring grooves.
  • Black color for visual identification.
  • Suitable for high-temperature laboratory sealing applications.
  • Used to seal reaction vessels and lids.