Learn More
Chemglass Life Sciences Simriz 486 FFKM O-Ring, AS568-110 (ID 9.20 mm x 2.62 mm)
SDP
Catalog No. 502743508
Encompass_Preferred
Perfluoroelastomer (FFKM) O-ring molded from the Simriz 486 compound, intended for demanding chemical and thermal sealing applications such as pharmaceutical and semiconductor equipment. The material provides high chemical resistance and a firm sealing profile in a white finish.
  • Perfluoroelastomer construction for high chemical resistance.
  • 75 Shore A durometer for sealing resilience.
  • Inner diameter 9.20 mm and cross section 2.62 mm.
  • White finish suitable for cleanroom and pharmaceutical environments.
  • Material datasheet available for performance and compatibility data.
Catalog No. 50-274-3508 Supplier Chemglass Life Sciences Supplier No. CG309Z110
Perfluoroelastomer (FFKM) O-ring molded from the Simriz 486 compound, intended for demanding chemical and thermal sealing applications such as pharmaceutical and semiconductor equipment. The material provides high chemical resistance and a firm sealing profile in a white finish.
  • Perfluoroelastomer construction for high chemical resistance.
  • 75 Shore A durometer for sealing resilience.
  • Inner diameter 9.20 mm and cross section 2.62 mm.
  • White finish suitable for cleanroom and pharmaceutical environments.
  • Material datasheet available for performance and compatibility data.