Learn More
Chemglass Life Sciences Chemglass O-Ring CG-309-Z-111 - Simriz 486 FFKM, AS568-111, white, USP Class VI
SDP
Catalog No. 502743507
Encompass_Preferred
The Chemglass O-ring CG-309-Z-111 is a white perfluoroelastomer (FFKM) seal made from Simriz 486 and certified to USP Class VI. It is designed for high-purity sealing in aggressive dry-process environments and is commonly used in semiconductor, pharmaceutical, and biotech applications.
  • Perfluoroelastomer construction for broad chemical resistance.
  • White color for high-purity applications and visual inspection.
  • USP Class VI certified for pharmaceutical and biotech use.
  • Suitable for aggressive dry-process chemistries and reactive plasmas.
  • Available in a wide range of AS568 standard sizes; consult manufacturer for specific size availability.
Catalog No. 50-274-3507 Supplier Chemglass Life Sciences Supplier No. CG309Z111
The Chemglass O-ring CG-309-Z-111 is a white perfluoroelastomer (FFKM) seal made from Simriz 486 and certified to USP Class VI. It is designed for high-purity sealing in aggressive dry-process environments and is commonly used in semiconductor, pharmaceutical, and biotech applications.
  • Perfluoroelastomer construction for broad chemical resistance.
  • White color for high-purity applications and visual inspection.
  • USP Class VI certified for pharmaceutical and biotech use.
  • Suitable for aggressive dry-process chemistries and reactive plasmas.
  • Available in a wide range of AS568 standard sizes; consult manufacturer for specific size availability.