Learn More
Chemglass Life Sciences Chemglass O-Ring #113 - Simriz 486 FFKM perfluoro (white), USP Class VI
SDP
Catalog No. 502743506
Encompass_Preferred
Chemglass O-Ring #113 is a white perfluoroelastomer (FFKM) sealing ring made from Simriz 486 designed for high-purity, chemically aggressive, and high-temperature environments. It retains elastomeric sealing properties while offering broad chemical resistance and is certified to USP Class VI.
  • Perfluoroelastomer (FFKM) construction for broad chemical resistance and elevated temperature performance.
  • White, high-purity formulation suitable for contamination-sensitive processes.
  • Certified to USP Class VI for biocompatibility and purity.
  • Suitable for aggressive chemistries and dry-process applications, including reactive plasmas.
  • Provides reliable elastomeric sealing where fluoropolymer-like chemical resistance is required.
Catalog No. 50-274-3506 Supplier Chemglass Life Sciences Supplier No. CG309Z113
Chemglass O-Ring #113 is a white perfluoroelastomer (FFKM) sealing ring made from Simriz 486 designed for high-purity, chemically aggressive, and high-temperature environments. It retains elastomeric sealing properties while offering broad chemical resistance and is certified to USP Class VI.
  • Perfluoroelastomer (FFKM) construction for broad chemical resistance and elevated temperature performance.
  • White, high-purity formulation suitable for contamination-sensitive processes.
  • Certified to USP Class VI for biocompatibility and purity.
  • Suitable for aggressive chemistries and dry-process applications, including reactive plasmas.
  • Provides reliable elastomeric sealing where fluoropolymer-like chemical resistance is required.