Learn More
Chemglass Life Sciences O-Ring AS568-115, Simriz 485 FFKM Perfluoroelastomer (Black) - For Laboratory Glassware Seals
SDP
Catalog No. 502743601
Encompass_Preferred
O-Ring AS568-115 made from Simriz 485 FFKM perfluoroelastomer. Designed for laboratory sealing applications that require broad chemical resistance and high thermal stability. Suitable for replacement seals in glassware fittings and process equipment.
  • Perfluoroelastomer offers broad chemical resistance.
  • Operates across a wide temperature range (-7°C to 230°C).
  • Shore A hardness approximately 75 for resilient sealing.
  • High tensile strength supports durability under stress.
  • Available in black color for easy visual identification.
Catalog No. 50-274-3601 Supplier Chemglass Life Sciences Supplier No. CG309B115
O-Ring AS568-115 made from Simriz 485 FFKM perfluoroelastomer. Designed for laboratory sealing applications that require broad chemical resistance and high thermal stability. Suitable for replacement seals in glassware fittings and process equipment.
  • Perfluoroelastomer offers broad chemical resistance.
  • Operates across a wide temperature range (-7°C to 230°C).
  • Shore A hardness approximately 75 for resilient sealing.
  • High tensile strength supports durability under stress.
  • Available in black color for easy visual identification.