Learn More
Chemglass Life Sciences Chemglass O-Ring, #116, Simriz 485 FFKM, Black
SDP
Catalog No. 502743600
Encompass_Preferred
Perfluoroelastomer O-ring designed for chemical resistance and high-temperature sealing in laboratory and fluid-handling equipment. Size #116 with 75 Shore A hardness and broad compatibility with aggressive chemicals.
  • Provides broad chemical resistance suitable for aggressive fluids.
  • Operates across a wide temperature range.
  • 75 Shore A hardness for general-purpose sealing.
  • High tensile strength for durable performance.
  • Sold individually for easy replacement.
Catalog No. 50-274-3600 Supplier Chemglass Life Sciences Supplier No. CG309B116
Perfluoroelastomer O-ring designed for chemical resistance and high-temperature sealing in laboratory and fluid-handling equipment. Size #116 with 75 Shore A hardness and broad compatibility with aggressive chemicals.
  • Provides broad chemical resistance suitable for aggressive fluids.
  • Operates across a wide temperature range.
  • 75 Shore A hardness for general-purpose sealing.
  • High tensile strength for durable performance.
  • Sold individually for easy replacement.