Learn More
Chemglass Life Sciences Perfluoroelastomer (FFKM) O-ring, AS568 #116, White, USP Class VI - for O-ring joints and seals in semiconductor and pharmaceutical equipment.
SDP
Catalog No. 502743504
Encompass_Preferred
High-purity perfluoroelastomer (FFKM) O-ring in AS568 size #116, white, and USP Class VI certified. It provides exceptional chemical resistance and wide temperature capability for demanding semiconductor, pharmaceutical, and biotechnology sealing applications.
  • High chemical resistance across a broad range of solvents and aggressive process chemistries.
  • Wide operating temperature range: -7°C to 230°C.
  • Shore A hardness of 75 for durable sealing performance.
  • Tensile strength approximately 2,620 psi for mechanical durability.
  • USP Class VI certification for biocompatibility and low contamination.
  • Manufactured in ISO-certified clean rooms to minimize particulates.
  • Suitable for vacuum joints, O-ring seals, and reactive plasma environments.
Catalog No. 50-274-3504 Supplier Chemglass Life Sciences Supplier No. CG309Z116
High-purity perfluoroelastomer (FFKM) O-ring in AS568 size #116, white, and USP Class VI certified. It provides exceptional chemical resistance and wide temperature capability for demanding semiconductor, pharmaceutical, and biotechnology sealing applications.
  • High chemical resistance across a broad range of solvents and aggressive process chemistries.
  • Wide operating temperature range: -7°C to 230°C.
  • Shore A hardness of 75 for durable sealing performance.
  • Tensile strength approximately 2,620 psi for mechanical durability.
  • USP Class VI certification for biocompatibility and low contamination.
  • Manufactured in ISO-certified clean rooms to minimize particulates.
  • Suitable for vacuum joints, O-ring seals, and reactive plasma environments.