Learn More
Chemglass Life Sciences Chemglass Perfluoro (FFKM) O-Ring, Simriz 486, Size #117 - seal for glassware joints and vacuum traps
SDP
Catalog No. 502743503
Encompass_Preferred
Chemglass perfluoro (FFKM) O-ring in size #117 made from a high-purity perfluoroelastomer. It is designed to provide broad chemical resistance and biocompatibility for sealing laboratory glassware joints, vacuum traps, and reaction vessel lids.
  • Perfluoro (FFKM) material provides broad chemical resistance.
  • White color enables visual inspection.
  • Class VI biocompatibility for pharmaceutical and semiconductor use.
  • 75 durometer hardness for reliable sealing performance.
  • Suitable for vacuum and high-purity laboratory applications.
Catalog No. 50-274-3503 Supplier Chemglass Life Sciences Supplier No. CG309Z117
Chemglass perfluoro (FFKM) O-ring in size #117 made from a high-purity perfluoroelastomer. It is designed to provide broad chemical resistance and biocompatibility for sealing laboratory glassware joints, vacuum traps, and reaction vessel lids.
  • Perfluoro (FFKM) material provides broad chemical resistance.
  • White color enables visual inspection.
  • Class VI biocompatibility for pharmaceutical and semiconductor use.
  • 75 durometer hardness for reliable sealing performance.
  • Suitable for vacuum and high-purity laboratory applications.