Learn More
Chemglass Life Sciences Chemglass O-Ring CG-309-Z-119 - Simriz 486 FFKM Perfluoro, Size 119, White, USP Class VI - For vacuum traps and O-ring joints
SDP
Catalog No. 502743502
Encompass_Preferred
These perfluoroelastomer (FFKM) O-rings use Simriz 486 material to provide broad chemical resistance and high temperature stability for demanding laboratory and semiconductor applications. They are produced in white color and are certified to USP Class VI for biocompatibility.
  • Perfluoroelastomer (FFKM) construction for broad chemical compatibility and plasma resistance.
  • White color and USP Class VI certification for biocompatibility.
  • Hardness, Shore A: 75.
  • Temperature range: -7°C to +230°C.
  • Tensile strength: 2,620 psi.
  • Suitable for vacuum traps and glassware O-ring joints.
Catalog No. 50-274-3502 Supplier Chemglass Life Sciences Supplier No. CG309Z119
These perfluoroelastomer (FFKM) O-rings use Simriz 486 material to provide broad chemical resistance and high temperature stability for demanding laboratory and semiconductor applications. They are produced in white color and are certified to USP Class VI for biocompatibility.
  • Perfluoroelastomer (FFKM) construction for broad chemical compatibility and plasma resistance.
  • White color and USP Class VI certification for biocompatibility.
  • Hardness, Shore A: 75.
  • Temperature range: -7°C to +230°C.
  • Tensile strength: 2,620 psi.
  • Suitable for vacuum traps and glassware O-ring joints.