Learn More
Chemglass Life Sciences Chemglass O-ring #120, FFKM perfluoroelastomer (white), USP Class VI
SDP
Catalog No. 502743501
Encompass_Preferred
O-ring size #120 made from a perfluoroelastomer (FFKM) compound engineered for broad chemical resistance and high-temperature stability. USP Class VI certification and a high-purity white compound make it suitable for pharmaceutical and semiconductor sealing applications.
  • Perfluoroelastomer (FFKM) construction for broad chemical and solvent resistance.
  • White, high-purity compound certified to USP Class VI.
  • Operating temperature range -7°C to 230°C.
  • Suitable for aggressive dry-process chemistries, including reactive plasmas.
  • Available in standard O-ring size #120 for replacement and maintenance.
Catalog No. 50-274-3501 Supplier Chemglass Life Sciences Supplier No. CG309Z120
O-ring size #120 made from a perfluoroelastomer (FFKM) compound engineered for broad chemical resistance and high-temperature stability. USP Class VI certification and a high-purity white compound make it suitable for pharmaceutical and semiconductor sealing applications.
  • Perfluoroelastomer (FFKM) construction for broad chemical and solvent resistance.
  • White, high-purity compound certified to USP Class VI.
  • Operating temperature range -7°C to 230°C.
  • Suitable for aggressive dry-process chemistries, including reactive plasmas.
  • Available in standard O-ring size #120 for replacement and maintenance.