Learn More
Chemglass Life Sciences Chemglass O-Ring #121, Simriz 486 FFKM (Perfluoroelastomer), White, USP Class VI
SDP
Catalog No. 502743500
Encompass_Preferred
Chemglass O-Ring #121 is molded from Simriz 486 perfluoroelastomer (FFKM) to provide high-purity, perfluoro sealing for chemically aggressive and high-temperature process environments. The white, USP Class VI-certified formulation offers broad chemical resistance and thermal stability for semiconductor, pharmaceutical, and other sensitive applications.
  • Perfluoroelastomer construction for broad chemical resistance.
  • High temperature resistance up to 220°C.
  • USP Class VI certified for biocompatibility.
  • White color suitable for ultra-clean processes.
  • Suitable for semiconductor, pharmaceutical, and biotech sealing applications.
  • Manufactured in certified clean-room conditions for consistent purity.
Catalog No. 50-274-3500 Supplier Chemglass Life Sciences Supplier No. CG309Z121
Chemglass O-Ring #121 is molded from Simriz 486 perfluoroelastomer (FFKM) to provide high-purity, perfluoro sealing for chemically aggressive and high-temperature process environments. The white, USP Class VI-certified formulation offers broad chemical resistance and thermal stability for semiconductor, pharmaceutical, and other sensitive applications.
  • Perfluoroelastomer construction for broad chemical resistance.
  • High temperature resistance up to 220°C.
  • USP Class VI certified for biocompatibility.
  • White color suitable for ultra-clean processes.
  • Suitable for semiconductor, pharmaceutical, and biotech sealing applications.
  • Manufactured in certified clean-room conditions for consistent purity.