Learn More
Chemglass Life Sciences O-Ring #122, Simriz 486 FFKM - white Class VI replacement for drain valves and reaction vessel lids
SDP
Catalog No. 502743499
Encompass_Preferred
Replacement perfluoroelastomer O-ring made from Simriz 486 FFKM. Size #122, white, and certified to Class VI. Intended as a spare sealing element for drain valves, stems, control knobs, and reaction vessel lids.
  • Perfluoroelastomer (FFKM) material for broad chemical resistance.
  • White color for visual identification.
  • Class VI biocompatibility certification.
  • Fits O-ring size number 122.
  • Used as a replacement sealing element for drain valves and reaction vessel lids.
Catalog No. 50-274-3499 Supplier Chemglass Life Sciences Supplier No. CG309Z122
Replacement perfluoroelastomer O-ring made from Simriz 486 FFKM. Size #122, white, and certified to Class VI. Intended as a spare sealing element for drain valves, stems, control knobs, and reaction vessel lids.
  • Perfluoroelastomer (FFKM) material for broad chemical resistance.
  • White color for visual identification.
  • Class VI biocompatibility certification.
  • Fits O-ring size number 122.
  • Used as a replacement sealing element for drain valves and reaction vessel lids.