Learn More
Chemglass Life Sciences Chemglass O-Ring #129 for AS568 lab joints and vacuum traps, Perfluoro (FFKM), white, Class VI
SDP
Catalog No. 502743498
Encompass_Preferred
Chemglass O-Ring #129 is a perfluoro (FFKM) sealing ring manufactured for high-purity laboratory and process applications. The compound delivers broad chemical resistance and elevated temperature performance suitable for vacuum, pharmaceutical, and semiconductor uses.
  • Perfluoroelastomer (FFKM) material for broad chemical compatibility.
  • White color and Class VI cleanliness for high-purity environments.
  • Shore A hardness 75 for durable sealing under pressure.
  • Service temperature range from -7 °C to 230 °C.
  • Sized to industry-standard O-ring size 129 for AS568-compatible joints.
  • Suitable for laboratory glassware joints, vacuum traps, and high-purity process seals.
Catalog No. 50-274-3498 Supplier Chemglass Life Sciences Supplier No. CG309Z129
Chemglass O-Ring #129 is a perfluoro (FFKM) sealing ring manufactured for high-purity laboratory and process applications. The compound delivers broad chemical resistance and elevated temperature performance suitable for vacuum, pharmaceutical, and semiconductor uses.
  • Perfluoroelastomer (FFKM) material for broad chemical compatibility.
  • White color and Class VI cleanliness for high-purity environments.
  • Shore A hardness 75 for durable sealing under pressure.
  • Service temperature range from -7 °C to 230 °C.
  • Sized to industry-standard O-ring size 129 for AS568-compatible joints.
  • Suitable for laboratory glassware joints, vacuum traps, and high-purity process seals.