Learn More
Chemglass Life Sciences Chemglass O-Ring #132 for Laboratory Glassware Sealing - Simriz 486 FFKM, White, Class VI (CG-309-Z-132)
SDP
Catalog No. 502743497
Encompass_Preferred
Perfluoro (FFKM) O-ring size #132 designed for sealing laboratory glassware and vacuum fittings. Manufactured from a high-purity perfluoroelastomer formulated for broad chemical resistance and elevated temperature stability, this O-ring is suitable for reaction vessels, lids, and O-ring joints in demanding lab environments.
  • Perfluoroelastomer (FFKM) construction.
  • White color for contamination control.
  • 75 Shore A hardness.
  • Temperature range: -7°C to 230°C.
  • USP Class VI compatible.
  • Suitable for laboratory glassware seals and vacuum fittings.
Catalog No. 50-274-3497 Supplier Chemglass Life Sciences Supplier No. CG309Z132
Perfluoro (FFKM) O-ring size #132 designed for sealing laboratory glassware and vacuum fittings. Manufactured from a high-purity perfluoroelastomer formulated for broad chemical resistance and elevated temperature stability, this O-ring is suitable for reaction vessels, lids, and O-ring joints in demanding lab environments.
  • Perfluoroelastomer (FFKM) construction.
  • White color for contamination control.
  • 75 Shore A hardness.
  • Temperature range: -7°C to 230°C.
  • USP Class VI compatible.
  • Suitable for laboratory glassware seals and vacuum fittings.