Learn More
Chemglass Life Sciences Simriz 486 FFKM O-Ring, AS568-136 - White, 75A (For laboratory seals and O-ring joints)
SDP
Catalog No. 502743496
Encompass_Preferred
Simriz 486 FFKM O-ring is a high-purity perfluoroelastomer seal designed for aggressive chemical environments and elevated temperatures. Supplied by Chemglass, it combines elastomer resilience with chemical resistance similar to PTFE and is certified to USP Class VI for high-purity laboratory and process applications.
  • High chemical resistance for aggressive process chemistries.
  • Wide operating temperature range suitable for elevated temperatures and thermal cycles.
  • White, clean-room compatible material for high-purity applications.
  • 75 Shore A hardness for durable sealing and low compression set.
  • Size: 1.987 in inner diameter x 0.103 in cross section (50.47 mm x 2.62 mm).
Catalog No. 50-274-3496 Supplier Chemglass Life Sciences Supplier No. CG309Z136
Simriz 486 FFKM O-ring is a high-purity perfluoroelastomer seal designed for aggressive chemical environments and elevated temperatures. Supplied by Chemglass, it combines elastomer resilience with chemical resistance similar to PTFE and is certified to USP Class VI for high-purity laboratory and process applications.
  • High chemical resistance for aggressive process chemistries.
  • Wide operating temperature range suitable for elevated temperatures and thermal cycles.
  • White, clean-room compatible material for high-purity applications.
  • 75 Shore A hardness for durable sealing and low compression set.
  • Size: 1.987 in inner diameter x 0.103 in cross section (50.47 mm x 2.62 mm).