Learn More
Chemglass Life Sciences Chemglass O-Ring #137, Simriz 486 FFKM, White, Class VI
SDP
Catalog No. 502743495
Encompass_Preferred
Perfluoroelastomer O-ring designed for high-temperature and aggressive chemical environments. The component provides broad chemical resistance and stability for dry-process and reactive plasma applications.
  • Perfluoroelastomer (FFKM) material for broad chemical resistance.
  • Size: #137.
  • White color for high-purity applications.
  • Class VI biocompatibility certification.
  • Heat resistance up to 220 °C.
  • Suitable for aggressive dry-process chemistries and reactive plasma environments.
Catalog No. 50-274-3495 Supplier Chemglass Life Sciences Supplier No. CG309Z137
Perfluoroelastomer O-ring designed for high-temperature and aggressive chemical environments. The component provides broad chemical resistance and stability for dry-process and reactive plasma applications.
  • Perfluoroelastomer (FFKM) material for broad chemical resistance.
  • Size: #137.
  • White color for high-purity applications.
  • Class VI biocompatibility certification.
  • Heat resistance up to 220 °C.
  • Suitable for aggressive dry-process chemistries and reactive plasma environments.