Learn More
Chemglass Life Sciences O-Ring #201 for O-Ring Joints and Vacuum Traps - Perfluoroelastomer (FFKM), White
SDP
Catalog No. 502743494
Encompass_Preferred
Chemglass O-ring manufactured from Simriz 486 perfluoroelastomer (FFKM) provides high purity, broad chemical resistance, and elevated temperature stability for demanding laboratory and semiconductor processes.
  • High chemical resistance to aggressive solvents and process chemistries.
  • White, high-purity perfluoroelastomer suitable for clean-room applications.
  • Heat resistance up to 220 °C.
  • Certified USP Class VI and produced in ISO-certified clean rooms.
  • Commonly used with O-ring joints and vacuum trap seals.
Catalog No. 50-274-3494 Supplier Chemglass Life Sciences Supplier No. CG309Z201
Chemglass O-ring manufactured from Simriz 486 perfluoroelastomer (FFKM) provides high purity, broad chemical resistance, and elevated temperature stability for demanding laboratory and semiconductor processes.
  • High chemical resistance to aggressive solvents and process chemistries.
  • White, high-purity perfluoroelastomer suitable for clean-room applications.
  • Heat resistance up to 220 °C.
  • Certified USP Class VI and produced in ISO-certified clean rooms.
  • Commonly used with O-ring joints and vacuum trap seals.