Learn More
Chemglass Life Sciences Chemglass O-Ring, AS568 #204 - Perfluoroelastomer (Simriz 486, CG-309-Z-204)
SDP
Catalog No. 502743493
Encompass_Preferred
Chemglass perfluoro (FFKM) O-ring made from Simriz 486 material, intended for laboratory sealing applications that require broad chemical compatibility and elevated temperature resistance.
  • Perfluoroelastomer compound with broad chemical resistance.
  • White color for cleanroom and pharmaceutical applications.
  • 75 Shore A durometer for high temperature stability.
  • Sized to AS568 #204 for standard O-ring fittings.
  • Suitable for vacuum and harsh chemical environments.
Catalog No. 50-274-3493 Supplier Chemglass Life Sciences Supplier No. CG309Z204
Chemglass perfluoro (FFKM) O-ring made from Simriz 486 material, intended for laboratory sealing applications that require broad chemical compatibility and elevated temperature resistance.
  • Perfluoroelastomer compound with broad chemical resistance.
  • White color for cleanroom and pharmaceutical applications.
  • 75 Shore A durometer for high temperature stability.
  • Sized to AS568 #204 for standard O-ring fittings.
  • Suitable for vacuum and harsh chemical environments.