Learn More
Chemglass Life Sciences Chemglass O-Ring #205 - Simriz 486 FFKM (perfluoro), white, USP Class VI
SDP
Catalog No. 502743492
Encompass_Preferred
Chemglass O-Ring #205 is a perfluoroelastomer (FFKM) sealing ring manufactured from Simriz 486 compound. It offers high chemical and thermal resistance and is suitable for regulated applications such as pharmaceutical and semiconductor processing.
  • Perfluoroelastomer material for broad chemical resistance.
  • High thermal stability for continuous use to 230°C.
  • Certified to USP Class VI for regulated processes.
  • White color for easy visual inspection.
  • Hardness 75 Shore A and tensile strength approximately 2,620 psi.
  • Size designation #205 for compatible gland profiles.
Catalog No. 50-274-3492 Supplier Chemglass Life Sciences Supplier No. CG309Z205
Chemglass O-Ring #205 is a perfluoroelastomer (FFKM) sealing ring manufactured from Simriz 486 compound. It offers high chemical and thermal resistance and is suitable for regulated applications such as pharmaceutical and semiconductor processing.
  • Perfluoroelastomer material for broad chemical resistance.
  • High thermal stability for continuous use to 230°C.
  • Certified to USP Class VI for regulated processes.
  • White color for easy visual inspection.
  • Hardness 75 Shore A and tensile strength approximately 2,620 psi.
  • Size designation #205 for compatible gland profiles.