Learn More
Chemglass Life Sciences Chemglass O-ring #206 - Simriz 486 FFKM (white), USP Class VI, for reaction vessels and lids
SDP
Catalog No. 502743491
Encompass_Preferred
Chemglass O-Ring #206 in Simriz 486 FFKM is a high-purity perfluoroelastomer O-ring engineered for demanding chemical and temperature environments commonly encountered in pharmaceutical and semiconductor applications.
  • High-purity perfluoroelastomer material provides broad chemical resistance.
  • Certified to USP Class VI for biocompatibility and cleanliness.
  • Wide temperature range: -7°C to 230°C.
  • Manufactured in certified clean-room facilities to support critical processes.
  • 75 Shore A hardness with tensile strength approximately 2,620 psi.
  • Suitable for aggressive dry process chemistries, including reactive plasmas.
Catalog No. 50-274-3491 Supplier Chemglass Life Sciences Supplier No. CG309Z206
Chemglass O-Ring #206 in Simriz 486 FFKM is a high-purity perfluoroelastomer O-ring engineered for demanding chemical and temperature environments commonly encountered in pharmaceutical and semiconductor applications.
  • High-purity perfluoroelastomer material provides broad chemical resistance.
  • Certified to USP Class VI for biocompatibility and cleanliness.
  • Wide temperature range: -7°C to 230°C.
  • Manufactured in certified clean-room facilities to support critical processes.
  • 75 Shore A hardness with tensile strength approximately 2,620 psi.
  • Suitable for aggressive dry process chemistries, including reactive plasmas.