Learn More
Chemglass Life Sciences Chemglass O-Ring, perfluoro (FFKM) Simriz 486, size #208 - white USP Class VI seal for reaction vessels, lids, and vacuum joints
SDP
Catalog No. 502743490
Encompass_Preferred
Perfluoroelastomer (FFKM) O-ring, size #208, designed for high chemical resistance and elevated temperatures. USP Class VI compatible, white color, suitable for contamination-sensitive processes and aggressive dry chemistries such as plasma processing.
  • Perfluoroelastomer (FFKM) material provides broad chemical and solvent resistance.
  • White color suitable for contamination-sensitive applications.
  • Certified to USP Class VI for biocompatibility where required.
  • Temperature range suitable for elevated service: approximately -7°C to 230°C.
  • Hardness approximately 75 Shore A for durable sealing performance.
  • High tensile strength supports long service life in demanding environments.
  • Intended for use with reaction vessels, lids, vacuum traps, and similar laboratory glassware joints.
Catalog No. 50-274-3490 Supplier Chemglass Life Sciences Supplier No. CG309Z208
Perfluoroelastomer (FFKM) O-ring, size #208, designed for high chemical resistance and elevated temperatures. USP Class VI compatible, white color, suitable for contamination-sensitive processes and aggressive dry chemistries such as plasma processing.
  • Perfluoroelastomer (FFKM) material provides broad chemical and solvent resistance.
  • White color suitable for contamination-sensitive applications.
  • Certified to USP Class VI for biocompatibility where required.
  • Temperature range suitable for elevated service: approximately -7°C to 230°C.
  • Hardness approximately 75 Shore A for durable sealing performance.
  • High tensile strength supports long service life in demanding environments.
  • Intended for use with reaction vessels, lids, vacuum traps, and similar laboratory glassware joints.