Learn More
Chemglass Life Sciences Chemglass O-Ring #209, Simriz 485 FFKM Perfluoroelastomer (Black)
SDP
Catalog No. 502743598
Encompass_Preferred
The Chemglass O-Ring #209 (CG-309-B-209) is molded from Simriz 485 FFKM perfluoroelastomer to provide broad chemical compatibility and reliable performance at elevated temperatures. It is intended as a replacement or spare O-ring for laboratory sealing applications where aggressive chemicals and heat resistance are required.
  • Perfluoroelastomer material for broad chemical resistance.
  • High temperature compatibility: -7°C to 230°C.
  • Shore A hardness 75.
  • Tensile strength 2,690 psi.
  • Black color for visual identification.
  • General-purpose sealing applications in chemical equipment.
Catalog No. 50-274-3598 Supplier Chemglass Life Sciences Supplier No. CG309B209
The Chemglass O-Ring #209 (CG-309-B-209) is molded from Simriz 485 FFKM perfluoroelastomer to provide broad chemical compatibility and reliable performance at elevated temperatures. It is intended as a replacement or spare O-ring for laboratory sealing applications where aggressive chemicals and heat resistance are required.
  • Perfluoroelastomer material for broad chemical resistance.
  • High temperature compatibility: -7°C to 230°C.
  • Shore A hardness 75.
  • Tensile strength 2,690 psi.
  • Black color for visual identification.
  • General-purpose sealing applications in chemical equipment.