Learn More
Chemglass Life Sciences Chemglass O-Ring #209 - Perfluoro (FFKM), White, Class VI
SDP
Catalog No. 502743489
Encompass_Preferred
Perfluoroelastomer (FFKM) O-ring in white, designed for broad chemical resistance and high temperature performance. Certified for biocompatibility and produced to industry quality standards.
  • Perfluoroelastomer (FFKM) construction for broad chemical resistance.
  • Hardness Shore A 75.
  • Temperature range -7°C to +230°C.
  • Tensile strength 2,620 psi.
  • USP Class VI certified; manufactured to ISO/TS 16949, ISO 9001, and AS9100 standards.
  • Suitable for aggressive dry process chemistries and reactive plasma environments.
  • White color for high purity applications.
  • Fits standard #209 O-ring size.
Catalog No. 50-274-3489 Supplier Chemglass Life Sciences Supplier No. CG309Z209
Perfluoroelastomer (FFKM) O-ring in white, designed for broad chemical resistance and high temperature performance. Certified for biocompatibility and produced to industry quality standards.
  • Perfluoroelastomer (FFKM) construction for broad chemical resistance.
  • Hardness Shore A 75.
  • Temperature range -7°C to +230°C.
  • Tensile strength 2,620 psi.
  • USP Class VI certified; manufactured to ISO/TS 16949, ISO 9001, and AS9100 standards.
  • Suitable for aggressive dry process chemistries and reactive plasma environments.
  • White color for high purity applications.
  • Fits standard #209 O-ring size.