Learn More
Chemglass Life Sciences Chemglass O-Ring #210 for laboratory glassware - Simriz 485 FFKM, Black
SDP
Catalog No. 502743597
Encompass_Preferred
Chemglass O-Ring #210 is made from Simriz 485 FFKM perfluoroelastomer and provides broad chemical resistance and high-temperature performance for laboratory sealing applications. Designed for replacement and maintenance of laboratory glassware and equipment, it delivers durable mechanical properties and compatibility with aggressive chemicals.
  • Perfluoro FFKM material for broad chemical resistance.
  • High temperature capability: -7°C to 230°C.
  • Shore A 75 hardness with tensile strength 2,690 psi.
  • Sold individually for replacement or maintenance.
  • Suitable for sealing laboratory glassware and equipment.
Catalog No. 50-274-3597 Supplier Chemglass Life Sciences Supplier No. CG309B210
Chemglass O-Ring #210 is made from Simriz 485 FFKM perfluoroelastomer and provides broad chemical resistance and high-temperature performance for laboratory sealing applications. Designed for replacement and maintenance of laboratory glassware and equipment, it delivers durable mechanical properties and compatibility with aggressive chemicals.
  • Perfluoro FFKM material for broad chemical resistance.
  • High temperature capability: -7°C to 230°C.
  • Shore A 75 hardness with tensile strength 2,690 psi.
  • Sold individually for replacement or maintenance.
  • Suitable for sealing laboratory glassware and equipment.