Learn More
Chemglass Life Sciences Chemglass O-ring #210, Simriz 486 FFKM perfluoroelastomer (white)
SDP
Catalog No. 502743488
Encompass_Preferred
Perfluoroelastomer (FFKM) O-ring made from Simriz 486 material for high-purity sealing in demanding chemical, pharmaceutical, and semiconductor applications. Certified to USP Class VI and manufactured in accredited clean rooms to support contamination-sensitive processes.
  • High-purity perfluoroelastomer material suitable for aggressive chemistries.
  • White color for cleanroom and pharmaceutical compatibility.
  • Sized to standard #210 O-ring dimensions.
  • Wide service temperature range from -7°C to +230°C, with short-term heat resistance up to 220°C.
  • Broad resistance to chemicals and solvents.
  • Suitable for aggressive dry-process chemistries and reactive plasma environments.
  • Manufactured in accredited clean rooms meeting ISO/TS 16949, ISO 9001, and AS9100 standards.
Catalog No. 50-274-3488 Supplier Chemglass Life Sciences Supplier No. CG309Z210
Perfluoroelastomer (FFKM) O-ring made from Simriz 486 material for high-purity sealing in demanding chemical, pharmaceutical, and semiconductor applications. Certified to USP Class VI and manufactured in accredited clean rooms to support contamination-sensitive processes.
  • High-purity perfluoroelastomer material suitable for aggressive chemistries.
  • White color for cleanroom and pharmaceutical compatibility.
  • Sized to standard #210 O-ring dimensions.
  • Wide service temperature range from -7°C to +230°C, with short-term heat resistance up to 220°C.
  • Broad resistance to chemicals and solvents.
  • Suitable for aggressive dry-process chemistries and reactive plasma environments.
  • Manufactured in accredited clean rooms meeting ISO/TS 16949, ISO 9001, and AS9100 standards.