Learn More
Chemglass Life Sciences Chemglass O-Ring #212, Simriz 486 FFKM, White, USP Class VI - for vacuum traps and O-ring joints
SDP
Catalog No. 502743487
Encompass_Preferred
This size #212 O-ring is molded from a perfluoroelastomer designed for high chemical resistance and elevated temperature performance. It is white and certified to USP Class VI, making it suitable for demanding dry process chemistries and reactive plasma environments.
  • Perfluoroelastomer construction for broad chemical compatibility.
  • Certified to USP Class VI for high-purity applications.
  • Rated for elevated temperatures up to 220°C.
  • White color aids visual inspection in assemblies.
  • Suitable for sealing vacuum traps and O-ring joints in dry process systems.
Catalog No. 50-274-3487 Supplier Chemglass Life Sciences Supplier No. CG309Z212
This size #212 O-ring is molded from a perfluoroelastomer designed for high chemical resistance and elevated temperature performance. It is white and certified to USP Class VI, making it suitable for demanding dry process chemistries and reactive plasma environments.
  • Perfluoroelastomer construction for broad chemical compatibility.
  • Certified to USP Class VI for high-purity applications.
  • Rated for elevated temperatures up to 220°C.
  • White color aids visual inspection in assemblies.
  • Suitable for sealing vacuum traps and O-ring joints in dry process systems.