Learn More
Chemglass Life Sciences Chemglass O-Ring #213 - Perfluoro FFKM, Black - Replacement Seal for Lab Glassware and Fittings
SDP
Catalog No. 502743596
Encompass_Preferred
The Chemglass O-Ring #213 is a perfluoroelastomer (FFKM) sealing ring designed for demanding chemical and high-temperature applications. It provides reliable sealing performance for laboratory glassware and related fittings where broad chemical resistance is required.
  • Perfluoroelastomer (FFKM) material for broad chemical resistance.
  • Size number 213, suitable as a replacement O-ring for compatible lab fittings.
  • Hardness: 75 Shore A, offering good compression set resistance.
  • Temperature range: -7°C to 230°C for high-temperature applications.
  • Sold individually (each) for direct replacement use.
  • Suitable for aggressive acids, bases, and organic solvents.
Catalog No. 50-274-3596 Supplier Chemglass Life Sciences Supplier No. CG309B213
The Chemglass O-Ring #213 is a perfluoroelastomer (FFKM) sealing ring designed for demanding chemical and high-temperature applications. It provides reliable sealing performance for laboratory glassware and related fittings where broad chemical resistance is required.
  • Perfluoroelastomer (FFKM) material for broad chemical resistance.
  • Size number 213, suitable as a replacement O-ring for compatible lab fittings.
  • Hardness: 75 Shore A, offering good compression set resistance.
  • Temperature range: -7°C to 230°C for high-temperature applications.
  • Sold individually (each) for direct replacement use.
  • Suitable for aggressive acids, bases, and organic solvents.