Learn More
Chemglass Life Sciences Simriz 486 FFKM O-Ring, AS568-213 (23.39 x 3.53 mm)
SDP
Catalog No. 502743486
Encompass_Preferred
Simriz 486 FFKM O-ring in AS568-213 size is a perfluoroelastomer seal formulated for high-temperature and chemically aggressive environments. The compound delivers low particulation and strong plasma resistance for reliable sealing in dry-process and semiconductor applications.
  • Perfluoroelastomer material offers exceptional chemical and solvent resistance.
  • White color improves visual inspection during maintenance.
  • Maintains elasticity across a wide temperature range; heat resistance up to 220°C and low-temperature glass transition near -7°C.
  • Low particulation and plasma-resistant properties suitable for dry-process environments.
  • Standard O-ring cross-section dimensions for compatibility with common grooves and housings.
Catalog No. 50-274-3486 Supplier Chemglass Life Sciences Supplier No. CG309Z213
Simriz 486 FFKM O-ring in AS568-213 size is a perfluoroelastomer seal formulated for high-temperature and chemically aggressive environments. The compound delivers low particulation and strong plasma resistance for reliable sealing in dry-process and semiconductor applications.
  • Perfluoroelastomer material offers exceptional chemical and solvent resistance.
  • White color improves visual inspection during maintenance.
  • Maintains elasticity across a wide temperature range; heat resistance up to 220°C and low-temperature glass transition near -7°C.
  • Low particulation and plasma-resistant properties suitable for dry-process environments.
  • Standard O-ring cross-section dimensions for compatibility with common grooves and housings.