Learn More
Chemglass Life Sciences O-Ring size 217 (Simriz 486 FFKM), white, Class VI - for laboratory glassware joints and fittings
SDP
Catalog No. 502743485
Encompass_Preferred
Chemglass O-ring molded from Simriz 486 FFKM perfluoroelastomer for laboratory sealing applications. Designed with a Class VI material rating and white color for sensitive lab use.
  • Perfluoroelastomer (FFKM) construction for broad chemical and temperature resistance.
  • White color and Class VI rating suitable for sensitive laboratory applications.
  • Molded for consistent sealing performance in laboratory fittings and joints.
  • Durable and resilient for extended service in aggressive chemical environments.
Catalog No. 50-274-3485 Supplier Chemglass Life Sciences Supplier No. CG309Z217
Chemglass O-ring molded from Simriz 486 FFKM perfluoroelastomer for laboratory sealing applications. Designed with a Class VI material rating and white color for sensitive lab use.
  • Perfluoroelastomer (FFKM) construction for broad chemical and temperature resistance.
  • White color and Class VI rating suitable for sensitive laboratory applications.
  • Molded for consistent sealing performance in laboratory fittings and joints.
  • Durable and resilient for extended service in aggressive chemical environments.