Learn More
Chemglass Life Sciences O-Ring #218, Simriz 486 FFKM (White) for O-Ring Joints and Vacuum Traps
SDP
Catalog No. 502743484
Encompass_Preferred
Perfluoroelastomer O-ring made from Simriz 486 (FFKM) for high-purity laboratory and process applications. White, USP Class VI material provides broad chemical resistance and high temperature performance for demanding environments.
  • Made from Simriz 486 FFKM perfluoroelastomer.
  • White, USP Class VI high-purity material.
  • Provides broad chemical and solvent resistance.
  • Resistant to temperatures up to 220°C.
  • Suitable for pharmaceutical, semiconductor, and biotech applications.
  • Intended for use with O-ring joints and vacuum traps.
  • Manufactured in ISO-certified clean room facilities.
Catalog No. 50-274-3484 Supplier Chemglass Life Sciences Supplier No. CG309Z218
Perfluoroelastomer O-ring made from Simriz 486 (FFKM) for high-purity laboratory and process applications. White, USP Class VI material provides broad chemical resistance and high temperature performance for demanding environments.
  • Made from Simriz 486 FFKM perfluoroelastomer.
  • White, USP Class VI high-purity material.
  • Provides broad chemical and solvent resistance.
  • Resistant to temperatures up to 220°C.
  • Suitable for pharmaceutical, semiconductor, and biotech applications.
  • Intended for use with O-ring joints and vacuum traps.
  • Manufactured in ISO-certified clean room facilities.